• USA Home
  • P0650 - Poly(Glu, Lys) hydrobromide

EMAIL THIS PAGE TO A FRIEND
P0650 Sigma

Poly(Glu, Lys) hydrobromide

Glu:Lys (1:4), mol wt 150,000-300,000

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents, Polyamino Acids, Random Copolymers
feed ratio   Glu:Lys (1:4)
mol wt   mol wt 150,000-300,000
storage temp.   −20°C

Description

Analysis Note

Molecular weight based on viscosity.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Poly(Amino Acid) Block Copolymers for Drug Delivery and other Biomedical Applications

Carmen Scholz Department of Chemistry, University of Alabama in Huntsville Huntsville, AL 35899 Email: scholzc@uah.edu
Keywords: Addition reactions, Asymmetric synthesis, Building blocks, Catalysis, Degradations, Living polymerization, PEGylations, Polymerization reactions, Precipitation, Proteomics, Ring-opening polymerization, Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?