• USA Home
  • P0809 - Calmodulin from bovine brain

EMAIL THIS PAGE TO A FRIEND
P0809 Sigma

Calmodulin from bovine brain

crude

Synonym: CaM, Phosphodiesterase 3′,5′-cyclic nucleotide activator from bovine brain

Purchase

Properties

Related Categories Application Index, Biochemicals and Reagents, Cell Signaling Enzymes, Cyclic Nucleotide Metabolism, Enzymes, Inhibitors, and Substrates More...
quality   crude
storage temp.   −20°C
Gene Information   cow ... CALM3(520277)

Description

Biochem/physiol Actions

Ca2+ binding protein that is required for activation of cyclic nucleotide-dependent phosphodiesterase. It is also a cofactor/activator of nitric oxide synthase, calcineurin, and many kinases including ATPase, myosin light chain kinase, and CAM kinase I, II, and III. It mediates ryanodine receptor activation by cyclic ADP ribose and is involved in intracellular Ca2+ homeostasis.

Other Notes

A convenient source of low molecular weight acidic proteins

Physical form

Lyophilized powder containing calmodulin and S-100 proteins

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?