• USA Home
  • P0853 - Polygalacturonic acid potassium salt

EMAIL THIS PAGE TO A FRIEND
P0853 Sigma

Polygalacturonic acid potassium salt

from citrus fruit, ≥80% (titration)

Synonym: Polyanhydrogalacturonic acid, Potassium polypectate

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates P-Z, Polysaccharide More...
biological source   from citrus fruit
assay   ≥80% (titration)

Description

Biochem/physiol Actions

In land plants, polygalacturonic acids are major components of cell wall polysaccharides (pectins). Bound polygalacturonase-inhibiting peptides (PGIP) protect pectin from degradation by fungal polygalacturonases.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?