• USA Home
  • P0861 - Phospholipase A2 from porcine pancreas

EMAIL THIS PAGE TO A FRIEND
P0861 Sigma

Phospholipase A2 from porcine pancreas

≥10,000 unit/mL

Synonym: Lecithinase A, PLA2, Phosphatidylcholine 2-acylhydrolase

Purchase

Description

Biochem/physiol Actions

Hydrolyzes the β-ester bond of zwitterionic glycerophospholipids. Preferred substrates are phosphatidylcholine, phosphatidylethanolamine, and their plasmalogen analogues. Phosphatidylinositol and phosphatidylserine are also hydrolyzed. Aggressively attacks phospholipids in membranes of intact cells.

General description

A product of Novozyme Corp.

Unit Definition

One unit is equivalent to the amount of enzyme producing 1 μmole of free fatty acid per minute at pH 8 and 40 °C.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
SZ6114900

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?