• USA Home
  • P0991 - Simon Manostat® pump
EMAIL THIS PAGE TO A FRIEND
P0991 Sigma
Simon Manostat® pump
AC input 230 V AC (CEE 22 socket)
Purchase
Properties
Related Categories Labware, Peristaltic Pumps, Pumps More...
mfr. no.   Manostat
Description
General description
An all purpose analog pump designed to handle laboratory fluid transfer tasks. Economical and simple to operate, analog setting offers infinite flowrate adjustment. The chemically-resistant pump head (molded of glass filled Noryl®) has three stainless steel rollers and a Plexiglas safety cover.
• Standard Fluid Transfer: 2 year warranty
• Variable Flow from 1 mL/min to 3450 mL/min
• Analog single-turn speed control
• Lighted on/off switch. The on/off switch lets you maintain your flow rate setting when pump is turned off
• Reversible for purging of transfer lines
• 1/10 hp reversible motor has soft start and back EMF for 3% speed control
• Stackable painted steel housing is IP22
• 230 V models meet CE regulations for safety and noise immunity, 115 V models meet UL 508, IEC 1010 and cUL
All drives come complete with pump head and 4 ft silicone tubing (size 17). Start/stop operation by an open contact closure or open collector driver. Order foot switch model 73-750-000; (Remote models only). Operating temperature 0-40 °C.
Legal Information
Manostat is a registered trademark of Cole-Parmer Instrument Co.
Noryl is a registered trademark of Sabic Innovative Plastics IP B.V.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z375284 Masterflex pump drive, economy drive, AC input 190 - 260 V AC
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?