• USA Home
  • P0992 - Hanna precision pH meter
EMAIL THIS PAGE TO A FRIEND
P0992 Sigma
Hanna precision pH meter
Model pH 210
Purchase
Properties
Related Categories Benchtop, Hanna, Hanna pH Meters, Hanna pH Meters and Accessories, Labware,
feature   calibration manual one- or two-point; offset and slope calibration settings are displayed − an excellent teaching aid
  pH calibration automatic 1 or 2 points with 5 memorized buffers (pH 4.01, 6.86, 7.01, 9.18, 10.01)
  temp. compensation automatic (with probe HI 7669/2W) or manual from 0-100 °C
  temp. probe HI 7669/2W with 1 m cable included
  temperature compensation 0 ‑ 100 °C (manual)
mfr. no.   Model pH 210
measuring range   −1999-1999 mV voltage, resolution: 1 mV
  0-14 pH, resolution: 0.01
  0.0-100.0 °C temperature, accuracy: 0.5 °C, resolution: 0.1 °C
  0.00-14.00 pH, accuracy: 0.01, resolution: 0.01 pH unit
L × W × H   240 mm × 182 mm × 74 mm
Description
General description
Ergonomic and a large horizontal display showing both pH (or mV) and temperature at the same time. Calibration reduced to a few steps, all guided by the meter′s microprocessor. Stability indicator to ensure subsequent measurements are correct.
HI 1131B pH electrode, combination glass, interchangeable, BNC connector, 1 m cable included
Includes a 12 VDC adaptor, for Europe this will be fitted with a 2-pin Europlug.
Product does not include the electrode arm pictured.
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z673811 Hanna HI 2210 & HI 2211 bench pH meters, HI 2210, AC input 110 V
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?