• USA Home
  • P1075 - Mitex® PTFE filter membranes
EMAIL THIS PAGE TO A FRIEND
P1075 Sigma
Mitex® PTFE filter membranes
pore size 5.0 μm, diam. 47 mm
Purchase
Properties
Related Categories Filtration Supplies, Labware, Manufacturer Browser, Membrane Filters, Millipore,
mfr. no.   Millipore, LSWP04700
  Millipore, LSWP04700
diam.   47 mm
pore size   5.0 μm pore size
Description
General description
Unlaminated and hydrophobic, pre-wet with e.g. methanol before use with aqueous solutions
Legal Information
Mitex is a registered trademark of Merck KGaA, Darmstadt, Germany
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?