• USA Home
  • P1079 - Polyphenon 100 from green tea

EMAIL THIS PAGE TO A FRIEND
P1079 Sigma

Polyphenon 100 from green tea

Purchase

Properties

storage temp.   2-8°C

Description

Other Notes

Extract containing mixture of polyphenolic compounds

Quality

Minimum 80% total catechins

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

P1204 Polyphenon 60 from green tea

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?