• USA Home
  • P1106 - Anti-Potassium Channel TASK-2 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
P1106 Sigma

Anti-Potassium Channel TASK-2 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Ion Channels, Antibodies to Potassium Channels,
species reactivity   rat
application(s)   immunohistochemistry: suitable using rat kidney and pancreas sections
  western blot: 1:200 using rat kidney membranes
clone   polyclonal
antibody form   affinity isolated antibody
form   lyophilized powder
storage temp.   −20°C
Gene Information   human ... KCNK5(8645)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 483-499 of human TASK-2 (with additional N-terminal cysteine).

Physical form

Lyophilized at ~0.3 mg/ml from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, 5% sucrose, and 0.025% sodium azide.

Price and Availability


Antibody Explorer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?