• USA Home
  • P1114 - Pipettor Clamp-ette
EMAIL THIS PAGE TO A FRIEND
P1114 Sigma
Pipettor Clamp-ette
assorted colors
Purchase
Properties
Related Categories Labware, Liquid Handling Equipment, Pipettes, Pipettors and Tips, Pipettor Accessories, Pipettors More...
mfr. no.   Heathrow Scientific
color   assorted colors
Description
General description
Holds most major brands of Pipettors. This unique product keeps pipettors at your fingertips. It secures to table tops and benches with a few turns of a large plastic screw. It moves easily from one location to another, wherever you need it in the lab. Single clamp comes in blue; pack of 3 includes one blue, one red and one purple.
Legal Information
Clamp-ette is a trademark of Heathrow Scientific
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?