• USA Home
  • P1225 - L-α-Phosphatidyl-D-myo-inositol 3,4-diphosphate, dipalmitoyl

EMAIL THIS PAGE TO A FRIEND
P1225 Sigma

L-α-Phosphatidyl-D-myo-inositol 3,4-diphosphate, dipalmitoyl

  • Empirical Formula (Hill Notation) C41H81O19P3

  • Molecular Weight 970.99

  •  MDL number MFCD01861926

  •  PubChem Substance ID 329820081

Purchase

Properties

InChI Key   HKWJHKSHEWVOSS-OMDJCSNQSA-N
solubility   DMSO: soluble
  chloroform: soluble
  ethanol: soluble
storage temp.   −20°C

Description

Biochem/physiol Actions

Activates Ca2+-insensitive PKC isotypes δ, ε, and η; activates Akt (a serine-threonine kinase also known as PKBα) by direct interaction with the Akt pleckstrin homology domain.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P6990 1,2-Dipalmitoylphosphatidylinositol 3,4-diphosphate sodium salt, ≥98%, solid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?