• USA Home
  • P1242 - Hanna precision pH meter
EMAIL THIS PAGE TO A FRIEND
P1242 Sigma
Hanna precision pH meter
Model pH 212
Purchase
Properties
Related Categories Benchtop, Hanna, Hanna pH Meters, Hanna pH Meters and Accessories, Labware,
feature   calibration manual one- or two-point; offset and slope calibration settings are displayed − an excellent teaching aid
  temp. compensation automatic (with probe HI 7669/2W) or manual from 0-100 °C
  temperature compensation 0 ‑ 100 °C (manual)
mfr. no.   Model pH 212
measuring range   −1999-1999 mV voltage, resolution: 1 mV
  -2.000-16.000 pH, accuracy: 0.01-0.002, resolution: 0.01-0.001 pH unit
  0-14 pH, resolution: 0.01
  0.0-100.0 °C temperature, accuracy: 0.5 °C, resolution: 0.1 °C
L × W × H   240 mm × 182 mm × 74 mm
Description
General description
Selectable resolutions between 0.01 and 0.001 over a pH range of -2 to 16 designed for added accuracy. Large horizontal display shows both pH (or mV) and temperature at the same time. A stability indicator ensures a proper calibration.
HI 1131B pH electrode, combination, glass, interchangeable, 1 m cable included. pH calibration: automatic 1 or 2 points with 5 memorized buffers (pH 4.01, 6.86, 7.01, 9.18, 10.01)
Includes a 12 VDC adaptor, for Europe this will be fitted with a 2-pin Europlug.
Product does not include the electrode arm pictured.
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?