• USA Home
  • P1302 - Potassium chloride solution

EMAIL THIS PAGE TO A FRIEND
P1302 Sigma

Potassium chloride solution

20%

  • CAS Number 7447-40-7

  • Linear Formula KCl

  • Molecular Weight 74.55

  •  Beilstein Registry Number 1711999

  •  MDL number MFCD00011360

Purchase

Properties

Related Categories Chemical Synthesis, Inorganic Salts, Potassium, Synthetic Reagents
InChI Key   WCUXLLCKKVVCTQ-UHFFFAOYSA-M
concentration   20%

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?