• USA Home
  • P1312 - Phosphodiesterase 3′:5′-cyclic-nucleotide activator (calmodulin)−gold labeled 20 nm Colloidal Gold Labeled

EMAIL THIS PAGE TO A FRIEND
P1312 Sigma

Phosphodiesterase 3′:5′-cyclic-nucleotide activator (calmodulin)−gold labeled 20 nm Colloidal Gold Labeled

5.0 A520 units/mL, aqueous glycerol suspension

Purchase

Properties

form   aqueous glycerol suspension
concentration   5.0 A520 units/mL
storage temp.   −20°C

Description

Physical form

Suspension in approx. 50% glycerol containing 0.15 M NaCl, 0.01 M BES, pH 7.0, 0.02% PEG 20 and 0.02% sodium azide

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?