• USA Home
  • P1325 - Immobilon®-CD charge-modified PVDF Membranes
EMAIL THIS PAGE TO A FRIEND
P1325 Sigma
Immobilon®-CD charge-modified PVDF Membranes
Purchase
Properties
mfr. no.   Millipore, ICDM10100
size   10 cm × 10 cm
pore size   0.1 μm
Description
General description
Charge-modified (cationic) PVDF for use in internal sequencing and for high-yield protein recovery
Legal Information
Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?