• USA Home
  • P1361 - Excel Scientific serological pipettes
EMAIL THIS PAGE TO A FRIEND
P1361 Sigma
Excel Scientific serological pipettes
size 1.0 mL, standard tip, sterile
Purchase
Properties
Related Categories Excel Scientific, Excel Scientific Serological Pipettes, Labware, Liquid Handling Equipment, Manufacturer Browser,
description   individually wrapped
sterility   sterile
mfr. no.   Excel Scientific SER-0010-S01
size   1.0 mL
Description
General description
These novel tips yield faster flow rates and reduce cell shearing. Pipettes manufactured from virgin polystyrene, certified pyrogen-free and non-cytotoxic.
• Unique smooth one-piece construction eliminates sample hang-up and accidental breakage.
• Standard 5, 10, and 25 mL pipettes are two inches shorter than most, for ease of use in benchtop hoods, yet long and narrow enough to reach corners of all flasks.
• Superior accuracy: ±1.5%.
• Selection of tip styles for samples of different viscosities and shear sensitivities.
• Standard sterile packaging is one-side-paper one-side-plastic, static-free, easy to open.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?