• USA Home
  • P1367 - Hanna precision pH meter
EMAIL THIS PAGE TO A FRIEND
P1367 Sigma
Hanna precision pH meter
Model pH 213
Purchase
Properties
Related Categories Benchtop, Hanna, Hanna pH Meters, Hanna pH Meters and Accessories, Labware,
feature   calibration manual one- or two-point; offset and slope calibration settings are displayed − an excellent teaching aid
  pH calibration Automatic 1 or 2 points with 5 memorized buffers (pH 4.01, 6.86, 7.01, 9.18, 10.01)
  temp. compensation Automatic (with probe HI 7669/2W) or manual from 0-100 °C
  temp. probe HI 7669/2W with 1 m cable included.
  temperature compensation 0 ‑ 100 °C (manual)
mfr. no.   Model pH 213
measuring range   −1999-1999 mV voltage, resolution: 1 mV
  -2.00-16.00 pH, accuracy: 0.01 pH unit, resolution: 0.01-0.001 pH unit
  0-100 °C temperature, accuracy: 0.5 °C, resolution: 0.1 °C
  0-14 pH, resolution: 0.01
  ±999.9 mV mV1, accuracy: 0.05 mV, resolution: 0.1 mV, (ISE)
  ±999.9 mV mV2, accuracy: 0.05 mV, resolution: 0.01 mV, (ORP)
L × W × H   240 mm × 182 mm × 74 mm
Description
General description
HI 1131B pH electrode, combination, glass, interchangeable, BNC connector, 1 m cable included.
Similar to the pH 212 model, but it also measures mV in the range of ±999.9 mV when used with ISE and ORP electrodes.
Includes a 12 VDC adaptor, for Europe this will be fitted with a 2-pin Europlug.
Product does not include the electrode arm pictured.
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?