• USA Home
  • P138 - PPHT-fluorescein acetate

EMAIL THIS PAGE TO A FRIEND
P138 Sigma

PPHT-fluorescein acetate

solid

Synonym: (±)-2-(N-Fluoresceinylthioureidophenethyl-N-propyl)amino-5-hydroxyTetralin® acetic acid

  • Empirical Formula (Hill Notation) C42H39N3O6S · C2H4O2

  • Molecular Weight 773.89

  •  PubChem Substance ID 329820092

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Dopaminergics, Neuroscience, Neurotransmission,
InChI Key   KRYNVGVNRFYTEP-UHFFFAOYSA-N
form   solid
color   orange
solubility   DMSO: soluble1.6 mg/mL
  2-hydroxypropyl-β-cyclodextrin: insoluble
  H2O: insoluble

Description

Application

Fluorescein derivative of the D2 dopamine receptor agonist; useful in fluorescent labeling of D2 receptors.

Caution

Photosensitive

Legal Information

Tetralin is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?