• USA Home
  • P1398 - 5β-Pregnane-3β,11β,21-triol-20-one

EMAIL THIS PAGE TO A FRIEND
P1398 Sigma

5β-Pregnane-3β,11β,21-triol-20-one

Synonym: 3β,11β,21-Trihydroxy-5β-pregnan-20-one, 3β,5β-Tetrahydrocorticosterone

Purchase

Properties

form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?