EMAIL THIS PAGE TO A FRIEND
P140 Sigma

Pertussis toxin

Purchase

Properties

storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3462 6.1 / PGI
WGK Germany 
3
RTECS 
XW5883750

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P7208 Pertussis toxin from Bordetella pertussis, lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?