• USA Home
  • P1425 - d-Propoxyphene-d7 hydrochloride

EMAIL THIS PAGE TO A FRIEND
P1425 Sigma

d-Propoxyphene-d7 hydrochloride

Synonym: α-d-4-Dimethylamino-3-methyl-1-(phenyl-d5)-2-phenyl-2-(butyl-1,1-d2 propionate

Purchase

Properties

isotopic purity   98 atom % D
drug control   regulated under CDSA - not available from Sigma-Aldrich Canada

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?