• USA Home
  • P1463 - Potassium tetraborate tetrahydrate

EMAIL THIS PAGE TO A FRIEND
P1463 Sigma

Potassium tetraborate tetrahydrate

BioXtra, ≥99.5%

Synonym: Potassium biborate

Purchase

Properties

Related Categories Essential Chemicals, Materials Science, Metal and Ceramic Science, Potassium Salts, Research Essentials,
InChI Key   FZICZLCGJZRAHS-UHFFFAOYSA-N
product line   BioXtra
assay   ≥99.5%
impurities   <0.001% Phosphorus (P)
  <0.1% Insoluble matter
solubility   H2O: soluble1 M at 20 °C, clear, colorless
anion traces   chloride (Cl-): <0.05%
  sulfate (SO42-): <0.05%
cation traces   Al: <0.0005%
  Ca: <0.0005%
  Cu: <0.0005%
  Fe: <0.0005%
  Mg: <0.0005%
  NH4+: <0.05%
  Na: <0.05%
  Pb: <0.001%
  Zn: <0.0005%

Price and Availability


Ultra-High Purity Inorganics
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

P5754 Potassium tetraborate tetrahydrate, ReagentPlus®, ≥99.5%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?