• USA Home
  • P1471 - L-α-Phosphatidyl-D-myo-inositol 3,5-diphosphate, dioctanoyl

EMAIL THIS PAGE TO A FRIEND
P1471 Sigma

L-α-Phosphatidyl-D-myo-inositol 3,5-diphosphate, dioctanoyl

  • Empirical Formula (Hill Notation) C25H49O19P3

  • Molecular Weight 746.57

  •  MDL number MFCD02259163

  •  PubChem Substance ID 329820096

Purchase

Description

Biochem/physiol Actions

Component of lipid signaling pathway.

Price and Availability


The Future of Bioimaging

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?