EMAIL THIS PAGE TO A FRIEND
P1522 Sigma

Pharmalytes®

pH range 3.0 - 10.0

Synonym: Ampholytes

Purchase

Properties

Related Categories Capillary Electrophoresis, Capillary Isoelectric Focusing, Carrier Ampholytes, Isoelectric Focusing and Two Dimensional Electrophoresis, Molecular Biology,
sterility   sterile-filtered
pH range   3.0 - 10.0
capacity   0.35 mmol/pH·mL buffering capacity

Description

Other Notes

IEF carrier ampholytes

Application

Pharmalytes® has been added in agarose for the preparation of isoelectric focusing gels.
It has been used in the sample buffer solution in capillary isoelectric focusing.

General description

Ampholytes are used to generate a pH gradient under application of an electric field.

Legal Information

Pharmalyte is a registered trademark of GE Healthcare

Price and Availability


Amicon Pro Purification System
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?