• USA Home
  • P1540 - Pipette tips, 250 μL
EMAIL THIS PAGE TO A FRIEND
P1540 Sigma
Pipette tips, 250 μL
Calibrated
Purchase
Description
General description
Reference markings at 10, 20, 50, 100 and 200 μL give visual confirmation of proper pipet operation.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?