EMAIL THIS PAGE TO A FRIEND
P1568 Sigma
Protein Folding
  •  ISBN-10 0-7167-7027-X
  •  ISBN-13 978-0-7167-7027-5
Purchase
Properties
Related Categories Books and Software, Labware, Proteomics Books More...
publication info   T. Creighton, ed., W.H. Freeman, 1992, 547 pp., hard cover
mfr. no.   Freeman
Description
General description
This book presents key experiments and investigative techniques as well as the implications of recent findings for modern biotechnology and biomedicine. The protein folding problem refers to the difficulty in determining how and why a protein adopts a particular conformation, given that the number of possible conformations for an average-sized protein approaches 10 to the 270th power. This subject is summarized in chapters on the physical basis of stability of folded conformations, mutational analysis of folding and stability, folding pathways, and other avenues of exploration.
Price and Availability
Detect Your Unseen Protein
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?