• USA Home
  • P1611 - Bacteriological pipettes
EMAIL THIS PAGE TO A FRIEND
P1611 Sigma
Bacteriological pipettes
size 1.1 mL, std tip, sterile
Purchase
Properties
Related Categories Excel Scientific, Excel Scientific Serological Pipettes, Labware, Liquid Handling Equipment, Manufacturer Browser,
sterility   sterile
mfr. no.   Excel Scientific SER-0011-S 01
size   1.1 mL
Description
General description
Open-end and wide-tip pipets yield faster flow rates and reduce shearing for food, meat, cosmetics, oils, paint, sludge and cell-culture applications. The 1.1 mL and 2.2 mL bacteriological pipets meet the A.P.H.A. standards for examination of dairy products
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?