• USA Home
  • P1615 - Pepsinogen II from human gastric mucosa

EMAIL THIS PAGE TO A FRIEND
P1615 Sigma

Pepsinogen II from human gastric mucosa

Purchase

Properties

Related Categories Application Index, Biochemicals and Reagents, Enzymes, Inhibitors, and Substrates, Proteases, Proteases & Protein Sequencing More...
shipped in   wet ice
storage temp.   2-8°C
Gene Information   human ... PGC(5225)

Description

Biochem/physiol Actions

Used as a marker of gastric mucosa status.

Physical form

Liquid, 20 mM Tris-HCl, pH 8.0

Application

The ratio between serum levels of pepsinogens I and II can be determined to identify patients with high risk of gastric cancer.

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?