EMAIL THIS PAGE TO A FRIEND
P1642 Sigma

Lauryl palmitoleate

liquid

Synonym: Lauryl palmitoleate, Palmitoleic acid lauryl ester

Purchase

Properties

Related Categories Biochemicals and Reagents, Esters, Fatty Acids and conjugates, Fatty Acyls, Lipids,
InChI Key   CDXFYPWJGOODOG-SQFISAMPSA-N
form   liquid
storage temp.   −20°C

Description

Packaging

100 mg in ampule

Sealed ampule.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

94791 SnapIT Ampule Opener, Regular, for 1-2ml, 5-10ml, 10-15ml ampoules
43917 SnapIT Ampule Opener, large, for 5-10ml, 10-15ml, 20-25ml ampoules

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?