EMAIL THIS PAGE TO A FRIEND
P1647 Sigma

Pharmalytes®

pH range 2.5 - 5.0

Synonym: Ampholytes

Purchase

Properties

Related Categories Capillary Electrophoresis, Capillary Isoelectric Focusing, Carrier Ampholytes, Isoelectric Focusing and Two Dimensional Electrophoresis, Molecular Biology,
sterility   sterile-filtered
pH range   2.5 - 5.0
capacity   0.35 mmol/pH·mL buffering capacity

Description

Other Notes

IEF carrier ampholytes

Legal Information

Pharmalyte is a registered trademark of GE Healthcare

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?