• USA Home
  • P1662 - R-Phycoerythrin from Porphyra tenera “Nori”

EMAIL THIS PAGE TO A FRIEND
P1662 Sigma

R-Phycoerythrin from Porphyra tenera “Nori”

lyophilized powder

Synonym: PE

Purchase

Properties

form   lyophilized powder
composition   Protein, ~30% Lowry
storage temp.   2-8°C

Description

Packaging

Package size based on protein content

Physical form

Lyophilized powder containing sucrose, dithioerythritol and sodium azide as preservatives

Application

For labeling proteins to make fluorescent probes

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?