• USA Home
  • P1666 - Poly-L-lysine–Agarose

EMAIL THIS PAGE TO A FRIEND
P1666 Sigma

Poly-L-lysine–Agarose

mol wt 70,000-150,000, saline suspension

Purchase

Properties

Related Categories Affinity Chromatography, Amino Acid Resins, Molecular Biology, Protein Chromatography, Proteomics More...
form   saline suspension
mol wt   mol wt 70,000-150,000
extent of labeling   ≥4 mg per mL
matrix   cross-linked 4% beaded agarose
matrix activation   cyanogen bromide
matrix attachment   ε-amino
matrix spacer   1 atom
storage temp.   2-8°C

Description

Physical form

Suspension in 2.0 M NaCl containing 0.02% thimerosal

Price and Availability


Amplified Detection

Amicon Pro Purification System
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?