EMAIL THIS PAGE TO A FRIEND
P1731 Sigma
Powerpette® plus
mauve
Purchase
Properties
Related Categories Jencons, Labware, Liquid Handling Equipment, Manufacturer Browser, Pipette Controllers and Pumps,
material   plastic
  silicone
mfr. no.   Jencons
volume   1-100 mL
color   mauve
Description
General description
Powerpette Plus offers precision finger tip control in a streamlined pistol grip shape. Pipette holder is angled making it ideal for use within a laminar flow or biological safety cabinet. Fitted with 0.45 μm Filter and supplied complete with AC adaptor, two spare hydrophobic filters (1 × 0.45 μm and 1 × 0.2 μm), bench stand and instruction manual
Features:
•  8 hours battery charge life; nickel metal hydride, rechargeable during use
•  3 aspirate and dispense modes; all with variable speed including gravity dispense for TD pipettes
•  Suitable for all types of pipettes 1-100 mL
•  CE marked
Legal Information
Powerpette is a registered trademark of Jencons Scientific, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?