EMAIL THIS PAGE TO A FRIEND
P1738 Sigma

PARP-1 bovine

≥98% (SDS-PAGE), ≥300 U/mg, buffered aqueous glycerol solution

Synonym: NAD+ ADP-ribosyltransferase, Poly(ADP-ribose) Polymerase, Poly(ADP-ribose) Synthetase

Purchase

Properties

assay   ≥98% (SDS-PAGE)
form   buffered aqueous glycerol solution
storage temp.   −70°C
Gene Information   cow ... ADPRT(286764)

Description

Biochem/physiol Actions

PARP-1, a nuclear enzyme that synthesizes ADP-ribose polymers from NAD, specifically binds Zn2+ and DNA, and recognizes single-strand breaks in DNA. It is involved in base excision repair, both short-patch and long-patch, rejoining DNA strand breaks and plays a role in p53 expression and activation. A high level of basal neuronal DNA damage and PARP activity has been reported in rat brain tissue. PARP-1 was shown to be required for HIV-1 integration into DNA. If PARP-1 is deficient there is no productive HIV-1 infection.

Unit Definition

One unit synthesizes 1 nmol of poly(ADP-ribose)/min at pH 7.5 at 25 °C.

Physical form

Solution in 50 mM Tris-HCl, pH 7.4, containing 1 M NaCl and 10 mM β-mercaptoethanol.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?