• USA Home
  • P1740 - Protein Phosphatase 2A1 from rabbit muscle

EMAIL THIS PAGE TO A FRIEND
P1740 Sigma

Protein Phosphatase 2A1 from rabbit muscle

≥95%, ~2000 units/mg protein, solution

Synonym: PP2A1

Purchase

Properties

assay   ≥95%
form   solution
mol wt   mol wt 187 kDa (Based on subunit composition A.B.C2 (A= 60 kDa, B=55 kDa, C=36 kDa))
shipped in   dry ice
storage temp.   −20°C

Description

Biochem/physiol Actions

Protein phosphatase 2A1 is specific for phosphorylated serine and threonine residues, with a preference for sites in signal transduction kinases.

Unit Definition

One unit equals 1 nanomole of phosphate relased per minute from 15 mM phosphorylase A

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

P6993 Protein Phosphatase 2A1 bovine, ≥1500 units/mg protein

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?