• USA Home
  • P1894 - PTH-DL-cysteic acid sodium salt

EMAIL THIS PAGE TO A FRIEND
P1894 Sigma

PTH-DL-cysteic acid sodium salt

  • Linear Formula C10H9N2O4S2Na

  • Molecular Weight 308.31

  •  MDL number MFCD00070135

  •  PubChem Substance ID 24898254

Purchase

Properties

Related Categories Molecular Biology, Protein Sequencing, Protein Structural Analysis, Proteomics, Reagents for Protein Sequencing More...
InChI Key   GFWRUNFCIXFKOS-JZGIKJSDSA-N
storage temp.   −20°C

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?