EMAIL THIS PAGE TO A FRIEND
P1897 Sigma

Pharmalytes®

pH range 5.0 - 8.0

Synonym: Ampholytes

Purchase

Properties

Related Categories Capillary Electrophoresis, Capillary Isoelectric Focusing, Carrier Ampholytes, Isoelectric Focusing and Two Dimensional Electrophoresis, Molecular Biology,
sterility   sterile-filtered
pH range   5.0 - 8.0
capacity   0.35 mmol/pH·mL buffering capacity
storage temp.   2-8°C

Description

Other Notes

IEF carrier ampholytes

Legal Information

Pharmalyte is a registered trademark of GE Healthcare

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?