• USA Home
  • P1899 - Poly-N5-(3-hydroxypropyl)-L-glutamine

EMAIL THIS PAGE TO A FRIEND
P1899 Sigma

Poly-N5-(3-hydroxypropyl)-L-glutamine

Purchase

Properties

mol wt   mol wt 40,000-60,000
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?