EMAIL THIS PAGE TO A FRIEND
P1913 Sigma

Polymaleimide

mol wt 10,000-20,000

Purchase

Description

Analysis Note

Molecular weight determination based on the viscosity-mol. wt. relationship of poly(ethylene-co-maleic anhydride).

Price and Availability


Webinar: Recent Developments in RAFT Agents
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?