• USA Home
  • P1914 - Polysaccharide from Zoogloea ramigera

EMAIL THIS PAGE TO A FRIEND
P1914 Sigma

Polysaccharide from Zoogloea ramigera

Purchase

Properties

storage temp.   2-8°C

Description

Application

Precipitates heavy metals such as iron, copper and uranyl ions from aqueous solutions.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?