• USA Home
  • P1915 - Phosphodiesterase 3′:5′-Cyclic Nucleotide Activator from Porcine Brain

EMAIL THIS PAGE TO A FRIEND
P1915 Sigma

Phosphodiesterase 3′:5′-Cyclic Nucleotide Activator from Porcine Brain

>95% (SDS-PAGE), lyophilized powder, >40,000 units/mg protein (Lowry)

Purchase

Properties

assay   >95% (SDS-PAGE)
form   lyophilized powder
impurities   salt, essentially free
storage temp.   −20°C

Description

Unit Definition

One unit will stimulate 0.008 activated unit of phosphodiesterase, 3′:5′-cyclic nucleotide, P 0520, in a 3 ml reaction volume at pH 7.5 and 30°C, to 50% of the maximum activity of the enzyme when saturated with activator, in the presence of 0.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?