• USA Home
  • P1959 - Phosphatase, Alkaline−maleimide activated from calf intestine

EMAIL THIS PAGE TO A FRIEND
P1959 Sigma

Phosphatase, Alkaline−maleimide activated from calf intestine

lyophilized powder, ≥2,000 DEA units/mg protein

Purchase

Properties

Related Categories Alkaline Phosphatase, Application Index, Biochemicals and Reagents, Enzymes, Inhibitors, and Substrates, Immunolabeling More...
form   lyophilized powder
composition   Protein, ≥30% BCA
extent of labeling   4-8 mol maleimide per mol protein
storage temp.   −20°C

Description

Application

For direct conjugation to compounds containing free sulfhydryl groups for use as enzyme-linked detection reagent. Instructions for use included.

Other Notes

Alkaline phosphatase, from calf intestinal mucosa, activated with maleimidocaproic acid N-hydroxysuccinimide ester.

Packaging

Package size based on protein content

Unit Definition

One unit will hydrolyze 1.0 μmole of p-nitrophenyl phosphate per min at pH 9.8 at 37°C.

Physical form

Lyophilized powder containing trehalose and sodium phosphate

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?