• USA Home
  • P1985 - Platelet-Derived Growth Factor Soluble Receptor α human

EMAIL THIS PAGE TO A FRIEND
P1985 Sigma

Platelet-Derived Growth Factor Soluble Receptor α human

>97% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym: PDGF sRα

Purchase

Description

Analysis Note

The bioactivity is measured by its ability to inhibit human PDGF-AB or recombinant human PDGF-AA in quiescent NR6R-3T3 fibroblasts.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 2.5 mg bovine serum albumin

Biochem/physiol Actions

Recombinant human platelet-derived growth factor soluble receptor α, a member of the class III subfamily of receptor tyrosine kinases, binds all three PDGF isoforms with high affinity and induces receptor homo-and heterodimerization and signal transduction.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?