• USA Home
  • P2046 - Peptide Sequencing Standard

EMAIL THIS PAGE TO A FRIEND
P2046 Sigma

Peptide Sequencing Standard

lyophilized powder

Synonym: Tyr-Ala-Glu-Gly-Asp-Val-His-Ala-Thr-Ser-Lys-Pro-Ala-Arg-Arg

Purchase

Properties

Related Categories Molecular Biology, Protein Sequencing, Protein Structural Analysis, Proteomics, Standards for Protein Sequencing More...
InChI Key   JJNXAJMLWIIHBK-QOECIXTQSA-N
form   lyophilized powder
mol wt   average mol wt 1,658 Da by calculation
  monoisotopic mol wt 1,657 Da
packaging   vial of ~20 nmol
storage temp.   2-8°C

Description

Application

Useful in evaluating the efficiency of protein sequencers.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

B4186 Butyrylcholinesterase from human serum, vial of ≥4 units

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?