• USA Home
  • P2112 - Greiner PCR 384 well plates, polypropylene

EMAIL THIS PAGE TO A FRIEND
P2112 Sigma

Greiner PCR 384 well plates, polypropylene

384 well, full skirted

Synonym: 384 multiwell plates, 384 well microplates, 384 well microtiter plates, 384 well plates

Purchase

Properties

Related Categories Genomics, PCR and Arrays, Labware, Molecular Biology, Molecular Biology Supplies, PCR (Polymerase Chain Reaction) Supplies,
material   polypropylene
packaging   case of 60
mfr. no.   Greiner 785201
color   clear colorless plate
matrix active group   non-treated surface treatment

Description

General description

384W PCR microplate with 25 μL working volume and 45 μL total volume

Other Notes

384 well, FμLl-Skirt microplate, without lid, rounded square well design, blue alphanumeric well coding; length: 127.76 +/- 0,15 mm width: 85.48 +/- 0,15 mm curvature: approx.150 μm.

Suitability

MolecμLar Biology. Thin polypropylene design to support polymerase chain reactions (PCR). Free of detectable DNase/RNase, human DNA and pyrogens

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?