• USA Home
  • P2162 - Protamine sulfate salt from salmon

EMAIL THIS PAGE TO A FRIEND
P2162 Sigma

Protamine sulfate salt from salmon

amorphous powder, cell culture tested

Synonym: Salmine from salmon, Salmine

Purchase

Properties

form   amorphous powder
color   white
suitability   cell culture tested

Description

Application

May be used for the removal of DNA from protein samples or for the purification of DNA binding proteins. It may be used as an alternative to polybrene in retroviral-mediated gene transfer.

Protamine in the form of solid lipid nanoparticles (SLN) promoted transfection with plasmid DNA more efficiently and with less cytotoxicity than comparable SLNs composed of Esterquat-1.

Protamine in the form of solid lipid nanoparticles (SLN) promoted transfection with plasmid DNA more efficiently and with less cytotoxicity than comparable SLNs composed of Esterquat-1.

Biochem/physiol Actions

Protamine sulfate is a small cationic protein that binds and precipitates DNA. Neutralizes the anticoagulant activity of heparin. Inhibits lipoprotein lipase.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?