• USA Home
  • P2213 - Protein tyrosine kinase from bovine spleen

EMAIL THIS PAGE TO A FRIEND
P2213 Sigma

Protein tyrosine kinase from bovine spleen

buffered aqueous glycerol solution, ~0.02 units/μg protein (BCA)

Purchase

Properties

form   buffered aqueous glycerol solution
shipped in   dry ice
storage temp.   −20°C

Description

Application

A non-receptor-associated protein tyrosine kinase. Useful for the phosphorylation of tyrosine containing proteins.

Packaging

Package size based on protein content

Unit Definition

One unit will transfer 1.0 picomole of phosphate from ATP to a synthetic peptide (Biotin-KYEKIGEGTYGVVYK-amide) at pH 7.2 at 30°C. The peptide corresponds to the amino acids 6-20 of the cell division kinase p34cdc2.

Physical form

Solution containing 25 mM HEPES, pH 7.0, 1 mM EDTA, 1 mM DTT and 50% glycerol

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?