EMAIL THIS PAGE TO A FRIEND
P2223 Sigma

Phase Divider Gel

Heavy/15

Purchase

Description

Application

For organic extractions of nucleic acids. Forms a durable barrier between phases, facilitating decanting or pipetting into a fresh tube.

Features and Benefits

• Improve nucleic acid recovery 20-30%
• Reduce contamination from interphase
• Gel is inert; does not interfere with subsequent enzymatic reactions
• Convenient for samples from 10 μl to 20 ml

Packaging

The gel comes pre-dispensed in common sizes of centrifuge tubes. Just add your phenol or phenol/chloroform extraction mixture and spin. The gel migrates under centrifugal force, forming a barrier above the organic phase and interphase.

in 15 ml low-speed centrifuge tubes for 1-6 ml samples

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?