• USA Home
  • P2229 - Anti-Platelet-Derived Growth Factor Soluble Receptor β antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
P2229 Sigma

Anti-Platelet-Derived Growth Factor Soluble Receptor β antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym: Anti-PDGF Rβ

Purchase

Description

Immunogen

recombinant human soluble PDGF receptor β (PDGF Rβ) extracellular domain, expressed in NSO cells.

Physical form

Lyophilized from 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Specificity

The antibody will neutralize soluble PDGF Rβ mediated bioactivity. It shows less than 1% cross-reactivity with recombinant human PDGF sRα.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Documents

Certificate of Analysis

Certificate of Origin

Safety Information

WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?