EMAIL THIS PAGE TO A FRIEND
P2299 Sigma
Pipette tips
Purchase
Properties
material   polypropylene
mfr. no.   Finn
Description
General description
• Available in convenient hinged racked package
• Made of high-grade polypropylene to be fully autoclavable
• 8 × 12 tip configuration for multi-channel tip pipetting
Other Notes
The tips are packaged either loose in bags or in autoclavable trays which fully enclose the tips and maintain sterility. Tips are supplied loose in bags unless otherwise noted.
Eppendorf Biopur pipette tips are sterile and are certified DNase-, RNase-, ATP-, and pyrogen-free. Automated production and packaging prevent contamination from microorganisms and human contact. Molded from dye-free resins.
Eppendorf Envirotips pipette tips are dye-free tips racked in 8 × 12 compact plastic trays held in biodegradable, recyclable cardboard packaging, eliminating up to 60% of the plastic waste of conventional racks. The individual package consists of a double-sided box with a tray full of tips on each side. Tips can be used directly from this package or from an Envirobox.
Eppendorf Envirosets include a supply of Envirotips (quantity as indicated) plus a reusable, autoclavable Envirobox for sterilization or decontamination. The Envirobox has a sliding cover which allows row-by-row use (eight tips at a time) without exposing the entire tray.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
P3049 Finnpipette® pipette tips, Finntip 250 Universal, non-sterile
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?